(4-Benzyloxyphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-(Benzyloxy)phenyl)acetic acid

OR6813-25G
APOSOR6813-25GEA 56.7 EUR
APOSOR6813-25G
(4-Benzyloxyphenyl)essigsäure 98%
(4-Benzyloxyphenyl)essigsäure
Formel: (C₆H₅CH₂OC₆H₄)CH₂COOH
Molecular Weight: 242.27 g/mol
Schmelzpunkt: 119…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017540
CAS-Nummer: 6547-53-1
EINECS: 229-463-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR