(2,6-Dichlorphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichlorophenylacetic acid
OR6814-100G
OR6814-25G
OR6814-10G
APOSOR6814-100GEA
102
EUR
APOSOR6814-100G
APOSOR6814-25G
APOSOR6814-10G
(2,6-Dichlorphenyl)essigsäure 98%
(2,6-Dichlorphenyl)essigsäure
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 316 Schmelzpunkt: 158…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004320 CAS-Nummer: 6575-24-2 EINECS: 229-504-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...