(3-Chlor-4-hydroxyphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

OR6818-5G
APOSOR6818-5GEA 21.9 EUR
APOSOR6818-5G
(3-Chlor-4-hydroxyphenyl)essigsäure 98%
(3-Chlor-4-hydroxyphenyl)essigsäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004349
CAS-Nummer: 33697-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR