1-[2-(Methylsulphanyl)phenyl]piperazine 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-(Methylthio)phenyl]piperazine
OR6838-500MG
APOSOR6838-500MGEA
46.4
EUR
APOSOR6838-500MG
1-[2-(Methylsulphanyl)phenyl]piperazine 96%
1-[2-(Methylsulphanyl)phenyl]piperazine
Formel:
C₁₁H₁₆N₂S Molecular Weight: 208,33 g/mol Siedepunkt: 120…126 °C/0,2mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040795 CAS-Nummer: 1013-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...