1-Acetylindole-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Acetyl-3-indolecarboxaldehyde, 1-Acetyl-1H-indole-3-carbaldehyde

OR6847-1G OR6847-25G OR6847-5G
APOSOR6847-1GEA 21.9 EUR
APOSOR6847-1G APOSOR6847-25G APOSOR6847-5G
1-Acetylindole-3-carboxaldehyde
1-Acetylindole-3-carboxaldehyde
Formel: C₁₁H₉NO₂
Molecular Weight: 187.20 g/mol
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039691
CAS-Nummer: 22948-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR