1-(1-Phenylethyl)piperazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6850-1G
APOSOR6850-1GEA
0
EUR
APOSOR6850-1G
1-(1-Phenylethyl)piperazine 97%
1-(1-Phenylethyl)piperazine
Formel:
C₁₂H₁₈N₂ Siedepunkt: 91…94 °C (0,1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040738 CAS-Nummer: 69628-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...