Ethylpiperazin-1-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate

OR6852-500G
APOSOR6852-500GEA 185 EUR
APOSOR6852-500G
Ethylpiperazin-1-carboxylat 98%
Ethylpiperazin-1-carboxylat
Formel: C₇H₁₄N₂O₂
Molecular Weight: 158.2 g/mol
Siedepunkt: 272…274 °C
Dichte: 1.08 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005964
CAS-Nummer: 120-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR