2-(Dimethylamino)benzonitril 95+%

Lieferant: Apollo Scientific
Achtung

OR6854-1G OR6854-250MG
APOSOR6854-1GEA 102 EUR
APOSOR6854-1G APOSOR6854-250MG
2-(Dimethylamino)benzonitril 95+%
2-(Dimethylamino)benzonitril
Formel: C₉H₁₀N₂
Molecular Weight: 146.1891 g/mol
Siedepunkt: 130 °C/12mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD01313389
CAS-Nummer: 20925-24-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR