2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride

OR6859-1G OR6859-250MG
APOSOR6859-1GEA 21.9 EUR
APOSOR6859-1G APOSOR6859-250MG
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride
Formel: C₁₃H₁₆N₂O₃
Schmelzpunkt: 263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00729051
CAS-Nummer: 70918-74-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR