2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride
OR6859-1G
OR6859-250MG
APOSOR6859-1GEA
21.9
EUR
APOSOR6859-1G
APOSOR6859-250MG
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride 98%
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride
Formel:
C₁₃H₁₆N₂O₃ Schmelzpunkt: 263 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00729051 CAS-Nummer: 70918-74-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...