Sodium 3-bromopyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Sodium 3-bromopicolinate

OR6866T-100MG
APOSOR6866T-100MGEA 72.5 EUR
APOSOR6866T-100MG
Sodium 3-bromopyridine-2-carboxylate
Sodium 3-bromopyridine-2-carboxylate
Formel: C₆H₃BrNO₂·Na
Molecular Weight: 223,987 g/mol
Schmelzpunkt: 292…293 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD02683289
CAS-Nummer: 1189933-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR