1-(4,4'-Dichlorobenzhydryl)piperazine 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR6874-1G
OR6874-5G
OR6874-250MG
APOSOR6874-1GEA
43.5
EUR
APOSOR6874-1G
APOSOR6874-5G
APOSOR6874-250MG
1-(4,4'-Dichlorobenzhydryl)piperazine 98%
1-(4,4'-Dichlorobenzhydryl)piperazine
Formel:
C₁₇H₁₈Cl₂N₂ Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191212 CAS-Nummer: 27469-61-0 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...