Ethyl 4-(2-hydroxyethyl)piperazine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR6875-1G
APOSOR6875-1GEA 29 EUR
APOSOR6875-1G
Ethyl 4-(2-hydroxyethyl)piperazine-1-carboxylate 97%
Ethyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Formel: C₉H₁₈N₂O₃
Molecular Weight: 202,251 g/mol
Siedepunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191218
CAS-Nummer: 14000-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR