N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR6889-1G
APOSOR6889-1GEA
231
EUR
APOSOR6889-1G
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid 95+%
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid
Formel:
C₁₄H₁₅N₃O₂ Molecular Weight: 257,29 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203268 CAS-Nummer: 387350-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...