1-(2-Methoxyethyl)piperazin

Lieferant: Apollo Scientific

OR6898-5G
APOSOR6898-5GEA 26.1 EUR
APOSOR6898-5G
1-(2-Methoxyethyl)piperazin
1-(2-Methoxyethyl)piperazin
Formel: C₇H₁₆N₂O
Molecular Weight: 144,22 g/mol
MDL: MFCD00191214
CAS-Nummer: 13484-40-7
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR