1-Methylpiperidine-3-carboxylic acid hydrochloride 98%

Lieferant: Apollo Scientific
Gefahr

OR6929-10G
APOSOR6929-10GEA 127 EUR
APOSOR6929-10G
1-Methylpiperidine-3-carboxylic acid hydrochloride 98%
1-Methylpiperidine-3-carboxylic acid hydrochloride
Formel: C₇H₁₄ClNO₂
Schmelzpunkt: 180…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190236
CAS-Nummer: 19999-64-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR