1-Methylpiperidine-4-carboxylic acid hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

OR6930-1G
APOSOR6930-1GEA 21.9 EUR
APOSOR6930-1G
1-Methylpiperidine-4-carboxylic acid hydrochloride 98%
1-Methylpiperidine-4-carboxylic acid hydrochloride
Formel: C₇H₁₄ClNO₂
Schmelzpunkt: 225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190235
CAS-Nummer: 71985-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR