4-Chlor-1-methylpiperidin Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

OR6934-250G
APOSOR6934-250GEA 328 EUR
APOSOR6934-250G
4-Chlor-1-methylpiperidin Hydrochlorid 97%
4-Chlor-1-methylpiperidin Hydrochlorid
Formel: C₆H₁₃Cl₂N
Schmelzpunkt: 158…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012834
CAS-Nummer: 5382-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR