1-Chloro-2,4-dimethyl-5-nitrobenzene 95+%

Lieferant: Apollo Scientific

OR6964-250MG
APOSOR6964-250MGEA 190 EUR
APOSOR6964-250MG
1-Chloro-2,4-dimethyl-5-nitrobenzene 95+%
1-Chloro-2,4-dimethyl-5-nitrobenzene
Formel: C₈H₈ClNO₂
Molecular Weight: 185,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00547853
CAS-Nummer: 69383-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR