Methyl-3-formyl-4-hydroxybenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-formyl-4-hydroxybenzoate

OR6967-250MG
APOSOR6967-250MGEA 21.9 EUR
APOSOR6967-250MG
Methyl-3-formyl-4-hydroxybenzoate
Methyl-3-formyl-4-hydroxybenzoate
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 80…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01661473
CAS-Nummer: 24589-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR