5-Ethylcyclohexane-1,3-dione 98%

Lieferant: Apollo Scientific
Achtung

OR6970-1G OR6970-250MG OR6970-500MG
APOSOR6970-1GEA 277 EUR
APOSOR6970-1G APOSOR6970-250MG APOSOR6970-500MG
5-Ethylcyclohexane-1,3-dione 98%
5-Ethylcyclohexane-1,3-dione
Formel: C₈H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00129457
CAS-Nummer: 862168-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR