4-Acetamido-3-ethylbenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR6971-1G
APOSOR6971-1GEA
154
EUR
APOSOR6971-1G
4-Acetamido-3-ethylbenzonitrile
4-Acetamido-3-ethylbenzonitrile
Formel:
C₁₁H₁₂N₂O Molecular Weight: 188,226 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221461 CAS-Nummer: 34921-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...