Methyl 3-hydroxy-4-(hydroxymethyl)benzoate

Lieferant: Apollo Scientific
Achtung

OR6978-1G
APOSOR6978-1GEA 561 EUR
APOSOR6978-1G
Methyl 3-hydroxy-4-(hydroxymethyl)benzoate
Methyl 3-hydroxy-4-(hydroxymethyl)benzoate
Formel: C₉H₁₀O₄
Molecular Weight: 182,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445667
CAS-Nummer: 71780-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR