5-Bromo-2-(hydroxymethyl)phenol 95%

Lieferant: Apollo Scientific

OR6980-1G
APOSOR6980-1GEA 92.8 EUR
APOSOR6980-1G
5-Bromo-2-(hydroxymethyl)phenol 95%
5-Bromo-2-(hydroxymethyl)phenol
Formel: C₇H₇BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08445665
CAS-Nummer: 170434-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR