3-(4-Pyridyl)acrylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6984-25G
OR6984-5G
OR6984-1G
APOSOR6984-25GEA
94.4
EUR
APOSOR6984-25G
APOSOR6984-5G
APOSOR6984-1G
3-(4-Pyridyl)acrylic acid 97%
3-(4-Pyridyl)acrylic acid
Formel:
C₈H₇NO₂ Schmelzpunkt: 296 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040746 CAS-Nummer: 5337-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...