tert-Butyl-N-(Benzyloxy)carbamate 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR70016-25G
OR70016-5G
OR70016-100G
APOSOR70016-25GEA
59.6
EUR
APOSOR70016-25G
APOSOR70016-5G
APOSOR70016-100G
tert-Butyl-N-(Benzyloxy)carbamate 98%
tert-Butyl-N-(Benzyloxy)carbamate
Formel:
C₁₂H₁₇NO₃ Schmelzpunkt: 45…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191873 CAS-Nummer: 79722-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...