(Pyrrolidin-3-yl)acetic acid hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Carboxymethyl)pyrrolidine hydrochloride, 3-(Carboxymethyl)tetrahydropyrrole hydrochloride, 2-(Pyrrolidin-3-yl)acetic acid hydrochloride

OR70020-1G
APOSOR70020-1GEA 229 EUR
APOSOR70020-1G
(Pyrrolidin-3-yl)acetic acid hydrochloride 98%
(Pyrrolidin-3-yl)acetic acid hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795694
CAS-Nummer: 80616-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR