4-Ethoxy-3-nitroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-ethoxynitrobenzene, 4-Amino-2-nitrophenetole

OR70022-1G
APOSOR70022-1GEA 102 EUR
APOSOR70022-1G
4-Ethoxy-3-nitroaniline
4-Ethoxy-3-nitroaniline
Formel: C₈H₁₀N₂O₃
Molecular Weight: 182,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036065
CAS-Nummer: 1777-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR