Methyl-3-brom-1H-indazol-4-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-bromo-1H-indazole-4-carboxylate

OR70023-1G
APOSOR70023-1GEA 87 EUR
APOSOR70023-1G
Methyl-3-brom-1H-indazol-4-carboxylat
Methyl-3-brom-1H-indazol-4-carboxylat
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,0681 g/mol
Schmelzpunkt: 132…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840642
CAS-Nummer: 885271-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR