2,3-Dihydro-3,3-dimethyl-1,5-benzoxazepin-4(5H)-one

Lieferant: Apollo Scientific

Synonyme: 3,3-Dimethyl-4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepine

OR70024-1G
APOSOR70024-1GEA 545 EUR
APOSOR70024-1G
2,3-Dihydro-3,3-dimethyl-1,5-benzoxazepin-4(5H)-one
2,3-Dihydro-3,3-dimethyl-1,5-benzoxazepin-4(5H)-one
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19374936
CAS-Nummer: 1267772-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR