4-Bromo-2-(methylsulphonyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
OR70027-1G
OR70027-250MG
OR70027-5G
APOSOR70027-1GEA
0
EUR
APOSOR70027-1G
APOSOR70027-250MG
APOSOR70027-5G
4-Bromo-2-(methylsulphonyl)aniline
4-Bromo-2-(methylsulphonyl)aniline
Formel:
C₇H₈BrNO₂S Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD21193892 CAS-Nummer: 1396554-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...