Cyclopropylmethanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR70033-250MG
APOSOR70033-250MGEA 169 EUR
APOSOR70033-250MG
Cyclopropylmethanesulphonyl chloride
Cyclopropylmethanesulphonyl chloride
Formel: C₄H₇ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD10697744
CAS-Nummer: 114132-26-2
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR