2-Brom-3-nitrophenol 97+%

Lieferant: Apollo Scientific
Achtung

OR70053-1G OR70053-25G OR70053-5G
APOSOR70053-1GEA 45.1 EUR
APOSOR70053-1G APOSOR70053-25G APOSOR70053-5G
2-Brom-3-nitrophenol 97+%
2-Brom-3-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.0100 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08704555
CAS-Nummer: 101935-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR