5-Brom-2-methoxyzimtsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR7006-5G
OR7006-1G
OR7006-10G
APOSOR7006-5GEA
43.5
EUR
APOSOR7006-5G
APOSOR7006-1G
APOSOR7006-10G
5-Brom-2-methoxyzimtsäure 97%
5-Brom-2-methoxyzimtsäure
Formel:
C₁₀H₉BrO₃ Schmelzpunkt: 224…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017596 CAS-Nummer: 40803-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...