2-Biphenylyl[bis(2-methyl-2-propanyl)]phosphin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR70086-5G
OR70086-25G
OR70086-100G
APOSOR70086-5GEA
71.2
EUR
APOSOR70086-5G
APOSOR70086-25G
APOSOR70086-100G
2-Biphenylyl[bis(2-methyl-2-propanyl)]phosphin 97%
2-(Di-tert-butylphosphino)biphenyl
Formel:
C₂₀H₂₇P Schmelzpunkt: 84…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862440 CAS-Nummer: 224311-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...