Chlor(1,5-cyclooctadien)rhodium(I) dimer 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1,5-Cyclooctadiene)rhodium chloride dimer
OR70089-1G
OR70089-250MG
OR70089-100MG
APOSOR70089-1GEA
582
EUR
APOSOR70089-1G
APOSOR70089-250MG
APOSOR70089-100MG
Chlor(1,5-cyclooctadien)rhodium(I) dimer 98%
Chlor(1,5-cyclooctadien)rhodium(I) dimer
Formel:
C₁₆H₂₄Cl₂Rh₂ Molecular Weight: 493.08 g/mol Schmelzpunkt: 243 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012415 CAS-Nummer: 12092-47-6 EINECS: 235-157-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...