p-Hydrazinobenzenesulphonic acid hemihydrate
Lieferant: Apollo Scientific
|
Achtung
|
OR70100-25G
APOSOR70100-25GEA
98.6
EUR
APOSOR70100-25G
p-Hydrazinobenzenesulphonic acid hemihydrate
p-Hydrazinobenzenesulphonic acid hemihydrate
Formel:
C₆H₈N₂O₃S Schmelzpunkt: 286 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00480958 CAS-Nummer: 854689-07-9 EINECS: 202-694-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...