5-Brombenzimidazol 98%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-1H-benzimidazole
OR70114-25G
APOSOR70114-25GEA
72.5
EUR
APOSOR70114-25G
5-Brombenzimidazol 98%
5-Bromobenzimidazole
Formel:
C₇H₅BrN₂ Molecular Weight: 197.04 g/mol Schmelzpunkt: 130…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160001 CAS-Nummer: 4887-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...