2-Methyl-4-sulphamoylbenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Aminosulphonyl)-2-methylbenzeneboronic acid, 4-Borono-3-methylbenzenesulphonamide

OR70128-100MG
APOSOR70128-100MGEA 145 EUR
APOSOR70128-100MG
2-Methyl-4-sulphamoylbenzeneboronic acid
2-Methyl-4-sulphamoylbenzeneboronic acid
Formel: C₇H₁₀BNO₄S
Molecular Weight: 215,035 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19684107
CAS-Nummer: 1402238-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR