2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Aminosulphonyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)toluene, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

OR70129-100MG
APOSOR70129-100MGEA 128 EUR
APOSOR70129-100MG
2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester 95%
2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester
Formel: C₁₃H₂₀BNO₄S
Molecular Weight: 297.178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18732812
CAS-Nummer: 1261295-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR