5-Chinolincarbaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chinolincarboxaldehyd
OR70131-5G
APOSOR70131-5GEA
517
EUR
APOSOR70131-5G
5-Chinolincarbaldehyd 98%
5-Chinolincarbaldehyd
Formel:
C₁₀H₇NO Molecular Weight: 157,17 g/mol Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00805835 CAS-Nummer: 22934-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...