4-Isopropylbenzene-1,3-diol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1,3-Dihydroxy-4-(prop-2-yl)benzene, 4-Isopropylresorcinol
OR70132-25G
APOSOR70132-25GEA
179
EUR
APOSOR70132-25G
4-Isopropylbenzene-1,3-diol
4-Isopropylbenzene-1,3-diol
Formel:
C₉H₁₂O₂ Molecular Weight: 152.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663778 CAS-Nummer: 23504-03-2 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...