1-Oxoisoindoline-4-carboxylic acid 97%
Lieferant: Apollo Scientific
OR70147-1G
APOSOR70147-1GEA
0
EUR
APOSOR70147-1G
1-Oxoisoindoline-4-carboxylic acid 97%
1-Oxoisoindoline-4-carboxylic acid
Formel:
C₉H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18417299 CAS-Nummer: 1261740-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...