5-Amino-2-methoxybenzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-methoxybenzoic Acid

OR70155-5G
APOSOR70155-5GEA 111 EUR
APOSOR70155-5G
5-Amino-2-methoxybenzoesäure
5-Amino-2-methoxybenzoesäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Schmelzpunkt: 161…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09260887
CAS-Nummer: 3403-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR