1,4-Phenylenediacryloyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR70156-1G
OR70156-250MG
APOSOR70156-1GEA
0
EUR
APOSOR70156-1G
APOSOR70156-250MG
1,4-Phenylenediacryloyl chloride
1,4-Phenylenediacryloyl chloride
Formel:
C₁₂H₈Cl₂O₂ Schmelzpunkt: 171…172 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01109676 CAS-Nummer: 35288-49-4 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...