(3S)-3-Methylpiperazine hydrochloride, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: (S)-1-Boc-3-Methylpiperazine hydrochloride

OR70192-100G
APOSOR70192-100GEA 404 EUR
APOSOR70192-100G
(3S)-3-Methylpiperazine hydrochloride, N1-BOC protected
(3S)-3-Methylpiperazine hydrochloride, N1-BOC protected
Formel: C₁₀H₂₀N₂O₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD11101275
CAS-Nummer: 1353006-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR