4,6-Dichloropyrimidine-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-4,6-dichloropyrimidine

OR70201-1G
APOSOR70201-1GEA 655 EUR
APOSOR70201-1G
4,6-Dichloropyrimidine-2-carboxylic acid 95%
4,6-Dichloropyrimidine-2-carboxylic acid
Formel: C₅H₂Cl₂N₂O₂
Molecular Weight: 192,988 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09834770
CAS-Nummer: 684220-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR