2-Brom-4-chlor-6-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR7021-5G
OR7021-100G
OR7021-25G
APOSOR7021-5GEA
21.9
EUR
APOSOR7021-5G
APOSOR7021-100G
APOSOR7021-25G
2-Brom-4-chlor-6-nitroanilin
2-Brom-4-chlor-6-nitroanilin
Formel:
C₆H₄BrClN₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD08275709 CAS-Nummer: 827-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...