2-Brom-4-chlor-6-nitroanilin

Lieferant: Apollo Scientific
Achtung

OR7021-5G OR7021-100G OR7021-25G
APOSOR7021-5GEA 21.9 EUR
APOSOR7021-5G APOSOR7021-100G APOSOR7021-25G
2-Brom-4-chlor-6-nitroanilin
2-Brom-4-chlor-6-nitroanilin
Formel: C₆H₄BrClN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD08275709
CAS-Nummer: 827-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR