2-Brom-3-iodtoluol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-iodo-3-methylbenzene

OR70218-5G
APOSOR70218-5GEA 59.6 EUR
APOSOR70218-5G
2-Brom-3-iodtoluol 95%
2-Brom-3-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296.9310 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11977389
CAS-Nummer: 888214-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR